Thermo Scientific Chemicals

2,3,4,5,6-Pentafluorobenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog number: A11623.14
25 g, Each
Thermo Scientific Chemicals

2,3,4,5,6-Pentafluorobenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog number: A11623.14
25 g, Each
Quantity
Catalog number: A11623.14
also known as A11623-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1765-40-8
IUPAC Name
1-(bromomethyl)-2,3,4,5,6-pentafluorobenzene
Molecular Formula
C7H2BrF5
InChI Key
XDEPVFFKOVDUNO-UHFFFAOYSA-N
SMILES
FC1=C(F)C(F)=C(CBr)C(F)=C1F
Refractive Index
1.4695-1.4735 @ 20°C
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥96.0%
Form
Liquid
Identification (FTIR)
Conforms

Description

2,3,4,5,6-Pentafluorobenzyl bromide is used as a reagent for derivatization of alcohols, carboxylic acids, sulfonamides for GC and polyfunctional thiols. It is used for the preparation of pentafluorobenzyl esters of organic acids for determination by capillary and GC. It is also used to derivatize N-7-substituted guanine adducts of DNA for determination by GC-electron capture-MS.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,4,5,6-Pentafluorobenzyl bromide is used as a reagent for derivatization of alcohols, carboxylic acids, sulfonamides for GC and polyfunctional thiols. It is used for the preparation of pentafluorobenzyl esters of organic acids for determination by capillary and GC. It is also used to derivatize N-7-substituted guanine adducts of DNA for determination by GC-electron capture-MS.

Solubility
Hydrolyzes in water.

Notes
Store in a cool, dry, well-ventilated area. Incompatible with bases and oxidizing agents. Store at 4°C. Protected from moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text