Thermo Scientific Chemicals

2-Fluoropyridine, 99%, Thermo Scientific Chemicals

Catalog number: A11627.14
25 g, Each
Thermo Scientific Chemicals

2-Fluoropyridine, 99%, Thermo Scientific Chemicals

Catalog number: A11627.14
25 g, Each
Quantity
Catalog number: A11627.14
also known as A11627-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
372-48-5
IUPAC Name
2-fluoropyridine
Molecular Formula
C5H4FN
InChI Key
MTAODLNXWYIKSO-UHFFFAOYSA-N
SMILES
FC1=CC=CC=N1
Refractive Index
1.4655-1.4685 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Form
Liquid

Description

2-Fluoropyridine is a useful precursor of 2,3-disubstituted pyridines which is made by the possibility of metallation at C-3, together with the ready nucleophilic displacement of the activated fluorine. It can also react with thiophene to get 2-thiophen-2-yl-pyridine. It is used in the synthesis of aminopyridines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoropyridine is a useful precursor of 2,3-disubstituted pyridines which is made by the possibility of metallation at C-3, together with the ready nucleophilic displacement of the activated fluorine. It can also react with thiophene to get 2-thiophen-2-yl-pyridine. It is used in the synthesis of aminopyridines.

Solubility
Soluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text