Thermo Scientific Chemicals

1-Bromo-4-fluorobenzene, 99%, Thermo Scientific Chemicals

Catalog number: A11630.36
500 g, Each
Thermo Scientific Chemicals

1-Bromo-4-fluorobenzene, 99%, Thermo Scientific Chemicals

Catalog number: A11630.36
500 g, Each
Quantity
Catalog number: A11630.36
also known as A11630-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
460-00-4
IUPAC Name
1-bromo-4-fluorobenzene
Molecular Formula
C6H4BrF
InChI Key
AITNMTXHTIIIBB-UHFFFAOYSA-N
SMILES
FC1=CC=C(Br)C=C1
Assay (GC)
≥98.5%
Appearance (Color)
Clear, colourless
Refractive Index
1.5250-1.5280
Form
Liquid
Identification (FTIR)
Conforms

Description

1-Bromo-4-fluorobenzene is used as an intermediate in the synthesis of atypical antipsychotic agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-4-fluorobenzene is used as an intermediate in the synthesis of atypical antipsychotic agents.

Solubility
Soluble in water(0.14g/L).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text