Thermo Scientific Chemicals

3-(3,4-Dihydroxyphenyl)propionic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A11638.06
5 g, Each
Thermo Scientific Chemicals

3-(3,4-Dihydroxyphenyl)propionic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A11638.06
5 g, Each
Quantity
Catalog number: A11638.06
also known as A11638-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1078-61-1
IUPAC Name
3-(3,4-dihydroxyphenyl)propanoic acid
Molecular Formula
C9H10O4
InChI Key
DZAUWHJDUNRCTF-UHFFFAOYSA-N
SMILES
OC(=O)CCC1=CC=C(O)C(O)=C1
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
Pale cream to cream to brown or pale pink
Assay (Silylated GC)
≥98.0%

Description

3-(3,4-Dihydroxyphenyl)propionic acid it can be used to produce 3-(3,4-dihydroxy-phenyl)-propionic acid octyl ester by heating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(3,4-Dihydroxyphenyl)propionic acid it can be used to produce 3-(3,4-dihydroxy-phenyl)-propionic acid octyl ester by heating.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text