Thermo Scientific Chemicals

2-Bromo-2-nitropropane-1,3-diol, 98+%, Thermo Scientific Chemicals

Catalog number: A11639.22
100 g, Each
Thermo Scientific Chemicals

2-Bromo-2-nitropropane-1,3-diol, 98+%, Thermo Scientific Chemicals

Catalog number: A11639.22
100 g, Each
Quantity
Catalog number: A11639.22
also known as A11639-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
52-51-7
IUPAC Name
2-bromo-2-nitropropane-1,3-diol
Molecular Formula
C3H6BrNO4
InChI Key
LVDKZNITIUWNER-UHFFFAOYSA-N
SMILES
OCC(Br)(CO)[N+]([O-])=O
Appearance (Color)
White to yellow
Form
Crystals or powder or crystalline powder
Assay (Titration ex Bromide)
≥98.0 to ≤102.0%
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms

Description

2-Bromo-2-nitropropane-1,3-diol Cosmetics, Preservatives for milk samples, Topical medications and also provide wide spectrum of antibacterial activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-2-nitropropane-1,3-diol Cosmetics, Preservatives for milk samples, Topical medications and also provide wide spectrum of antibacterial activity.

Solubility
Soluble in water(280g/L).

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text