Thermo Scientific Chemicals

3-Amino-5-methyl-1H-pyrazole, 97%, Thermo Scientific Chemicals

Catalog number: A11642.14
25 g, Each
Thermo Scientific Chemicals

3-Amino-5-methyl-1H-pyrazole, 97%, Thermo Scientific Chemicals

Catalog number: A11642.14
25 g, Each
Quantity
Catalog number: A11642.14
also known as A11642-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
31230-17-8
IUPAC Name
5-methyl-1H-pyrazol-3-amine
Molecular Formula
C4H7N3
InChI Key
FYTLHYRDGXRYEY-UHFFFAOYSA-N
SMILES
CC1=CC(N)=NN1
Appearance (Color)
Pale yellow to orange or yellow-orange
Identification (FTIR)
Conforms
Form
Crystalline or fused solid
Assay (GC)
≥96.0%
Melting Point (clear melt)
39-47?C

Description

3-Amino-5-methyl-1H-pyrazole was employed as beta-sheet template to investigate its interaction with ferrocenoyl-dipeptides. It is also used in the synthesis of 4-aryl-3,7,7-trimethyl-2,4,6,7,8,9-hexahydropyrazolo[3,4-b]quinolin-5-ones. It can react with 4-Ethoxymethylene-2-phenyl-4H-oxazol-5-one to get N-(2-Methyl-7-oxo-4,7-dihydro-pyrazolo[1,5-a]pyrimidin-6-yl)-benzamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-5-methyl-1H-pyrazole was employed as beta-sheet template to investigate its interaction with ferrocenoyl-dipeptides. It is also used in the synthesis of 4-aryl-3,7,7-trimethyl-2,4,6,7,8,9-hexahydropyrazolo[3,4-b]quinolin-5-ones. It can react with 4-Ethoxymethylene-2-phenyl-4H-oxazol-5-one to get N-(2-Methyl-7-oxo-4,7-dihydro-pyrazolo[1,5-a]pyrimidin-6-yl)-benzamide.

Solubility
Soluble in Dichloromethane and Methanol.

Notes
Air Sensitive. Store at -20°C. Store under dry inert gas away from oxidizing agents and air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text