Thermo Scientific Chemicals

2-Octanone, 98%, Thermo Scientific Chemicals

Catalog number: A11643.0J
5000 mL, Each
Thermo Scientific Chemicals

2-Octanone, 98%, Thermo Scientific Chemicals

Catalog number: A11643.0J
5000 mL, Each
Quantity
Catalog number: A11643.0J
also known as A11643-0J
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-13-7
IUPAC Name
octan-2-one
Molecular Formula
C8H16O
InChI Key
ZPVFWPFBNIEHGJ-UHFFFAOYSA-N
SMILES
CCCCCCC(C)=O
Appearance (Color)
Clear colorless
Form
Liquid
Identification (FTIR)
Conforms
Refractive Index
1.4130-1.4170 @ 20?C
Assay (GC)
≥97.5%

Description

2-Octanone is used in perfumes, high-boiling solvent, especially for epoxy resin coatings and in leather finishes. It is used in flavoring, odorant. It can act as antiblushing agent for nitrocellulose lacquers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Octanone is used in perfumes, high-boiling solvent, especially for epoxy resin coatings and in leather finishes. It is used in flavoring, odorant. It can act as antiblushing agent for nitrocellulose lacquers.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text