Thermo Scientific Chemicals

Ethyl tetradecanoate, 98%, Thermo Scientific Chemicals

Catalog number: A11646.AP
500 mL, Each
Thermo Scientific Chemicals

Ethyl tetradecanoate, 98%, Thermo Scientific Chemicals

Catalog number: A11646.AP
500 mL, Each
Quantity
Catalog number: A11646.AP
also known as A11646-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
124-06-1
IUPAC Name
ethyl tetradecanoate
Molecular Formula
C16H32O2
InChI Key
MMKRHZKQPFCLLS-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCC(=O)OCC
Assay (GC)
≥97.5%
Form
Liquid
Refractive Index
1.4340-1.4380 @ 20?C
Appearance (Color)
Clear colorless

Description

A labeled component from essential oil of natural hawthorn perfume. Also frequently used in cosmetics, soaps, and flavorings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A labeled component from essential oil of natural hawthorn perfume. Also frequently used in cosmetics, soaps, and flavorings.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text