Thermo Scientific Chemicals

Sodium oxalate, 99%, Thermo Scientific Chemicals

Catalog number: A11648.0E
2500 g, Each
Thermo Scientific Chemicals

Sodium oxalate, 99%, Thermo Scientific Chemicals

Catalog number: A11648.0E
2500 g, Each
Quantity
Catalog number: A11648.0E
also known as A11648-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
62-76-0
IUPAC Name
disodium oxalate
Molecular Formula
C2Na2O4
InChI Key
ZNCPFRVNHGOPAG-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-]C(=O)C([O-])=O
Form
Powder
Appearance (Color)
White
Assay (Redox Titration)
≥98.5 to ≤101.5%
Water Content (Karl Fischer Titration)
<0.5%
Identification (FTIR)
Conforms

Description

Sodium oxalate is used as reducing agent, as a primary standard for standardizing potassium permanganate solutions and as a neutralizer for exothermic acidic reactions. It finds application in metal cleaning preparations, metal extraction and separation, blueprint coatings, electroplating baths, special cement manufacturing, plant nutrition as well as in textile and pharmaceutical industries. It plays an important role to remove calcium ions from blood plasma and prevents blood clotting.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium oxalate is used as reducing agent, as a primary standard for standardizing potassium permanganate solutions and as a neutralizer for exothermic acidic reactions. It finds application in metal cleaning preparations, metal extraction and separation, blueprint coatings, electroplating baths, special cement manufacturing, plant nutrition as well as in textile and pharmaceutical industries. It plays an important role to remove calcium ions from blood plasma and prevents blood clotting.

Solubility
Soluble in water and formic acid. Insoluble in ether and alcohol.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text