Thermo Scientific Chemicals

4-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: A11655.36
500 g, Each
Thermo Scientific Chemicals

4-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: A11655.36
500 g, Each
Quantity
Catalog number: A11655.36
also known as A11655-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
555-16-8
Identification (FTIR)
Conforms
Melting Point (clear melt)
102-108°C
Appearance (Color)
White to cream to yellow to orange
Form
Crystalline powder or powder
Assay (GC)
≥98.5%

Description

4-Nitrobenzaldehyde is used in the preparation of homoallylic alcohols. It is also involved in the development and evaluation of a series of tripeptide organocatalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrobenzaldehyde is used in the preparation of homoallylic alcohols. It is also involved in the development and evaluation of a series of tripeptide organocatalysts.

Solubility
Soluble in water, ethanol, benzene, glacial acetic acid. Slightly soluble in ether.

Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text