Thermo Scientific Chemicals

Potassium chloride, 99%, Thermo Scientific Chemicals

Catalog number: A11662.0B
1000 g, Each
Thermo Scientific Chemicals

Potassium chloride, 99%, Thermo Scientific Chemicals

Catalog number: A11662.0B
1000 g, Each
Quantity
Catalog number: A11662.0B
also known as A11662-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7447-40-7
IUPAC Name
potassium chloride
Molecular Formula
ClK
InChI Key
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES
[Cl-].[K+]
Appearance (Color)
White
Assay (Titration ex Chloride)
≥98.5% to ≤101.5%
Comment
Contains Anti Caking Agent (~0.8% magnesium hydroxide carbonate)
Form
Crystalline powder or powder or crystals

Description

The main use of potassium chloride is in the manufacture of fertilizers. It finds use in medicine as an electrolyte replenisher, and to treat hypokalemia with its pharmaceutical preparation, a condition in which the level of potassium will be low in blood. Further it finds use as a flux agent in welding and metal casting. It is useful in water softener units, certain de-icing products and calibration of the radiation monitoring equipment. It finds use as a scotophor in dark-trace CRTs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The main use of potassium chloride is in the manufacture of fertilizers. It finds use in medicine as an electrolyte replenisher, and to treat hypokalemia with its pharmaceutical preparation, a condition in which the level of potassium will be low in blood. Further it finds use as a flux agent in welding and metal casting. It is useful in water softener units, certain de-icing products and calibration of the radiation monitoring equipment. It finds use as a scotophor in dark-trace CRTs.

Solubility
Insoluble in water. Slightly soluble in alcohol. Insoluble in ether.

Notes
Hygroscopic.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text