3,5-Dihydroxybenzoic acid, 98%, Thermo Scientific Chemicals
3,5-Dihydroxybenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-Dihydroxybenzoic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number A11664.22
also known as A11664-22
Price (USD)/ Each
55.30
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
55.30
Add to cart
3,5-Dihydroxybenzoic acid, 98%, Thermo Scientific Chemicals
Catalog numberA11664.22
Price (USD)/ Each
55.30
-
Add to cart
Chemical Identifiers
CAS99-10-5
IUPAC Name3,5-dihydroxybenzoic acid
Molecular FormulaC7H6O4
InChI KeyUYEMGAFJOZZIFP-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(O)=CC(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale pink
Identification (FTIR)Conforms
FormPowder
Assay (HPLC)≥96.0%
Melting Point (clear melt)235-242?C
View more
3,5-Dihydroxybenzoic acid is used in organic synthesis and as intermediate for the pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dihydroxybenzoic acid is used in organic synthesis and as intermediate for the pharmaceutical.

Solubility
Soluble in 50 mg/mL, 95% Ethanol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Norbert Hoffmann.; Jean-Pierre Pete. Acid catalyzed intramolecular [2+2] photocycloaddition of 3,5-dihydroxybenzoic acid derivatives. Tetrahedron Letters. 1998, 39, (28), 5027-5030.