Thermo Scientific Chemicals

1H-Indazole, 99%, Thermo Scientific Chemicals

Catalog number: A11665.14
25 g, Each
Thermo Scientific Chemicals

1H-Indazole, 99%, Thermo Scientific Chemicals

Catalog number: A11665.14
25 g, Each
Quantity
Catalog number: A11665.14
also known as A11665-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
271-44-3
IUPAC Name
1H-indazole
Molecular Formula
C7H6N2
InChI Key
BAXOFTOLAUCFNW-UHFFFAOYSA-N
SMILES
N1N=CC2=CC=CC=C12
Assay (GC)
≥98.5%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
144.0-150.0?C
Appearance (Color)
White to cream
Identification (FTIR)
Conforms

Description

1H-Indazole is used in organic synthesis, it can react with butyryl chloride and 1-butyryl-1H-indazole. It is also used in synthesis of several small molecule inhibitors as potential cancer therapeutics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-Indazole is used in organic synthesis, it can react with butyryl chloride and 1-butyryl-1H-indazole. It is also used in synthesis of several small molecule inhibitors as potential cancer therapeutics.

Solubility
Soluble in hot water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text