Thermo Scientific Chemicals

1,4-Dibromobutane, 98+%, Thermo Scientific Chemicals

Catalog number: A11669.22
100 g, Each
Thermo Scientific Chemicals

1,4-Dibromobutane, 98+%, Thermo Scientific Chemicals

Catalog number: A11669.22
100 g, Each
Quantity
Catalog number: A11669.22
also known as A11669-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-52-1
IUPAC Name
1,4-dibromobutane
Molecular Formula
C4H8Br2
InChI Key
ULTHEAFYOOPTTB-UHFFFAOYSA-N
SMILES
BrCCCCBr
Identification (FTIR)
Conforms
Form
Liquid
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥98.0%
Refractive Index
1.5165-1.5205 @ 20?C

Description

1,4-Dibromobutane is used as an intermediate involved in the synthesis of active pharmaceutical ingredient and other organic compounds. It is also used in the investigation of metabolism of two halopropanes such as 1,3-dichloropropane and 2,2-dichloropropane. Further, it acts as a reagent to prepare diazadioxa oxovanadium(IV) macrocyclic complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dibromobutane is used as an intermediate involved in the synthesis of active pharmaceutical ingredient and other organic compounds. It is also used in the investigation of metabolism of two halopropanes such as 1,3-dichloropropane and 2,2-dichloropropane. Further, it acts as a reagent to prepare diazadioxa oxovanadium(IV) macrocyclic complexes.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text