Di-n-butylamine is an important intermediate used for various applications in organic synthesis. It is involved in the production of rubber chemicals like di-n-butylthiocarbamate thiuram, emulsifiers and dibutylethanolamine, which is used in coatings and paints. It is also used as plasticizer in PVC matrix and a corrosion inhibitor in an aqueous system. It plays an important role as a catalyst in vulcanization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Di-n-butylamine is an important intermediate used for various applications in organic synthesis. It is involved in the production of rubber chemicals like di-n-butylthiocarbamate thiuram, emulsifiers and dibutylethanolamine, which is used in coatings and paints. It is also used as plasticizer in PVC matrix and a corrosion inhibitor in an aqueous system. It plays an important role as a catalyst in vulcanization.
Solubility
Miscible with water, ether, ethanol and acetone.
Notes
Incompatible with strong oxidizing agents, carbon dioxide, zinc, iron and copper.
RUO – Research Use Only
General References:
- Oliveira, K. C. B.; Santos, A. G.; Santos, E. N. Hydroaminomethylation of eugenol with di-n-butylamine catalyzed by rhodium complexes: Bringing light on the promoting effect of Brönsted acids. Appl. Catal., A 2012, 445-446, 204-208.
- Chowdhury, F. I.; Saleh, M. A. Viscosities and deviations in viscosity of tert-butanol with n-butylamine, di-n-butylamine and tri-n-butylamine. J. Mol. Liq. 2014, 191, 156-160.