Thermo Scientific Chemicals

Dodecanoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A11672.0C
10,000 g, Each
Thermo Scientific Chemicals

Dodecanoic acid, 98%, Thermo Scientific Chemicals

Catalog number: A11672.0C
10,000 g, Each
Quantity
Catalog number: A11672.0C
also known as A11672-0C
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
143-07-7
IUPAC Name
dodecanoic acid
Molecular Formula
C12H24O2
InChI Key
POULHZVOKOAJMA-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCC(O)=O
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Melting Point (clear melt)
42.0-48.0?C
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder or flakes

Description

Dodecanoic acid is used in the preparation of cosmetics, soaps, alkyd resins and wetting agents. It is also used to measure the molar mass of an unknown substance through freezing point depression. It is also used as a food additive and an active component in a treatment for acne. In addition to this, it is a substrate for acylation of certain proteins based on the murine studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dodecanoic acid is used in the preparation of cosmetics, soaps, alkyd resins and wetting agents. It is also used to measure the molar mass of an unknown substance through freezing point depression. It is also used as a food additive and an active component in a treatment for acne. In addition to this, it is a substrate for acylation of certain proteins based on the murine studies.

Solubility
Soluble in water, benzene, acetone, alcohol, petroleum ether, dimethyl sulfoxide and dimethyl formamide. Slightly soluble in chloroform.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text