4-Methoxy-2-nitroaniline, 99%, Thermo Scientific Chemicals
4-Methoxy-2-nitroaniline, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methoxy-2-nitroaniline, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
100 g
500 g
Catalog number A11673.0E
also known as A11673-0E
Price (USD)/ Each
429.65
Online exclusive
477.00 
Save 47.35 (10%)
-
Add to cart
Quantity:
2500 g
Request bulk or custom format
Price (USD)/ Each
429.65
Online exclusive
477.00 
Save 47.35 (10%)
Add to cart
4-Methoxy-2-nitroaniline, 99%, Thermo Scientific Chemicals
Catalog numberA11673.0E
Price (USD)/ Each
429.65
Online exclusive
477.00 
Save 47.35 (10%)
-
Add to cart
Chemical Identifiers
CAS96-96-8
IUPAC Name4-methoxy-2-nitroaniline
Molecular FormulaC7H8N2O3
InChI KeyQFMJFXFXQAFGBO-UHFFFAOYSA-N
SMILESCOC1=CC=C(N)C(=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or lumps
Melting Point (clear melt)121.5-127.5?C
Appearance (Color)Orange to red
Assay (GC)≥98.5%
Solution Test(1% w/v in methanol) : Clear solution
4-Methoxy-2-nitroaniline is an aniline compound used in the synthesis of dyes and pigments. It has also been shown to act as a molluscicidal agent and has been used in hemolytic anemia studies. Also used as an intermediate for the synthesis of dyes, pigments and other chemical compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxy-2-nitroaniline is an aniline compound used in the synthesis of dyes and pigments. It has also been shown to act as a molluscicidal agent and has been used in hemolytic anemia studies. Also used as an intermediate for the synthesis of dyes, pigments and other chemical compounds.

Solubility
Slightly soluble in water. Soluble in hot methanol almost transparency.

Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. C. Ravikumara; and I. Hubert Joe. Electronic absorption and vibrational spectra and nonlinear optical properties of 4-methoxy-2-nitroaniline. Phys. Chem. Chem. Phys. 2010, 12 (32), 9452-9460.
  2. R.H. Contreras; D.G. De Kowalewski; J.C. Facelli .The NMR analysis of the methoxy-group conformation in 4-methoxy-2-nitroaniline. Journal of Molecular Structure. 1982, 81 (1-2), 147-149.