Thermo Scientific Chemicals

4-Methoxy-2-nitroaniline, 99%, Thermo Scientific Chemicals

Catalog number: A11673.22
100 g, Each
Thermo Scientific Chemicals

4-Methoxy-2-nitroaniline, 99%, Thermo Scientific Chemicals

Catalog number: A11673.22
100 g, Each
Quantity
Catalog number: A11673.22
also known as A11673-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
96-96-8
IUPAC Name
4-methoxy-2-nitroaniline
Molecular Formula
C7H8N2O3
InChI Key
QFMJFXFXQAFGBO-UHFFFAOYSA-N
SMILES
COC1=CC=C(N)C(=C1)[N+]([O-])=O
Form
Crystals or powder or crystalline powder or lumps
Melting Point (clear melt)
121.5-127.5?C
Appearance (Color)
Orange to red
Assay (GC)
≥98.5%
Solution Test
(1% w/v in methanol) : Clear solution

Description

4-Methoxy-2-nitroaniline is an aniline compound used in the synthesis of dyes and pigments. It has also been shown to act as a molluscicidal agent and has been used in hemolytic anemia studies. Also used as an intermediate for the synthesis of dyes, pigments and other chemical compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxy-2-nitroaniline is an aniline compound used in the synthesis of dyes and pigments. It has also been shown to act as a molluscicidal agent and has been used in hemolytic anemia studies. Also used as an intermediate for the synthesis of dyes, pigments and other chemical compounds.

Solubility
Slightly soluble in water. Soluble in hot methanol almost transparency.

Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text