1-Pentylamine, 98%, Thermo Scientific Chemicals
1-Pentylamine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Pentylamine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A11674.14
also known as A11674-14
Price (USD)/ Each
52.60
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
52.60
Add to cart
1-Pentylamine, 98%, Thermo Scientific Chemicals
Catalog numberA11674.14
Price (USD)/ Each
52.60
-
Add to cart
Chemical Identifiers
CAS110-58-7
IUPAC Namepentan-1-amine
Molecular FormulaC5H13N
InChI KeyDPBLXKKOBLCELK-UHFFFAOYSA-N
SMILESCCCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.4095-1.4135 @ 20?C
1-Pentylamine is used as a solvent, as a raw material in the manufacture of a variety of other compounds, including dyes, emulsifiers, and pharmaceutical products and as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pentylamine is used as a solvent, as a raw material in the manufacture of a variety of other compounds, including dyes, emulsifiers, and pharmaceutical products and as a flavoring agent.

Solubility
Soluble in alcohol, ether.

Notes
Air Sensitive. Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Patrik Spanel.; David Smith. Selected ion flow tube studies of the reactions of H3O+, NO+, and O2+ with eleven amine structural isomers of c5h13n1. International Journal of Mass Spectrometry. 1999, 185-187, 139-147.