Thermo Scientific Chemicals

1-Pentylamine, 98%, Thermo Scientific Chemicals

Catalog number: A11674.36
500 g, Each
Thermo Scientific Chemicals

1-Pentylamine, 98%, Thermo Scientific Chemicals

Catalog number: A11674.36
500 g, Each
Quantity
Catalog number: A11674.36
also known as A11674-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-58-7
IUPAC Name
pentan-1-amine
Molecular Formula
C5H13N
InChI Key
DPBLXKKOBLCELK-UHFFFAOYSA-N
SMILES
CCCCCN
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥97.5%
Form
Liquid
Refractive Index
1.4095-1.4135 @ 20?C

Description

1-Pentylamine is used as a solvent, as a raw material in the manufacture of a variety of other compounds, including dyes, emulsifiers, and pharmaceutical products and as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pentylamine is used as a solvent, as a raw material in the manufacture of a variety of other compounds, including dyes, emulsifiers, and pharmaceutical products and as a flavoring agent.

Solubility
Soluble in alcohol, ether.

Notes
Air Sensitive. Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text