Triethyloxonium tetrafluoroborate is used as an effective agent used for ethylation., laboratory chemicals, manufacture of substances.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Triethyloxonium tetrafluoroborate is used as an effective agent used for ethylation., laboratory chemicals, manufacture of substances.
Solubility
Soluble in Dichloromethane. Reacts with water.
Notes
Moisture Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only
General References:
- Douglas J. Raber.; Patrick GarianoJr.; Albert O. Brod.; Anne Gariano.; Wayne C. Guida.; Anne R. Guida.; Mark D. Herbst. Esterification of carboxylic acids with trialkyloxonium salts. J. Org. Chem. 1979, 44, (7), 1149-1154.
- Elena I. Klimova.; Tatiana Klimova Berestneva.; Simon Hernandez Ortega.; Daniel Mendez Iturbide.; Alfonso García Marquez.; Marcos Martínez García. Diferrocenylcyclopropenyl cations. Synthesis, structures, and some chemical properties. Journal of Organometallic Chemistry. 2005, 690, (14), 3333-3339.