Thermo Scientific Chemicals

Triethyloxonium tetrafluoroborate, 1.0M in dichloromethane, Thermo Scientific Chemicals

Catalog number: A11678.AP
500 mL, Each
Thermo Scientific Chemicals

Triethyloxonium tetrafluoroborate, 1.0M in dichloromethane, Thermo Scientific Chemicals

Catalog number: A11678.AP
500 mL, Each
Quantity
Catalog number: A11678.AP
also known as A11678-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
368-39-8
IUPAC Name
tetrafluoroboranuide; triethyloxidanium
Molecular Formula
C6H15BF4O
InChI Key
IYDQMLLDOVRSJJ-UHFFFAOYSA-N
SMILES
F[B-](F)(F)F.CC[O+](CC)CC
Comment
May form slight sediment on storage
Form
Liquid
Assay (Non-aqueous acid-base Titration)
1.0 ± 0.075M
Appearance (Color)
Clear or slightly turbid, colorless to pale yellow or pale pink

Description

Triethyloxonium tetrafluoroborate is used as an effective agent used for ethylation., laboratory chemicals, manufacture of substances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyloxonium tetrafluoroborate is used as an effective agent used for ethylation., laboratory chemicals, manufacture of substances.

Solubility
Soluble in Dichloromethane. Reacts with water.

Notes
Moisture Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text