Thermo Scientific Chemicals

2,4,6-Trichlorophenol, 98%, Thermo Scientific Chemicals

Catalog number: A11681.0B
1000 g, Each
Thermo Scientific Chemicals

2,4,6-Trichlorophenol, 98%, Thermo Scientific Chemicals

Catalog number: A11681.0B
1000 g, Each
Quantity
Catalog number: A11681.0B
also known as A11681-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
88-06-2
IUPAC Name
2,4,6-trichlorophenol
Molecular Formula
C6H3Cl3O
InChI Key
LINPIYWFGCPVIE-UHFFFAOYSA-N
SMILES
OC1=C(Cl)C=C(Cl)C=C1Cl
Assay (GC)
≥97.5%
Appearance (Color)
White to cream to yellow to pale brown
Form
Crystals or powder or crystalline powder or granules
Melting Point (clear melt)
62.0-69.0°C

Description

2,4,6-Trichlorophenol, antiseptic, defoliant, and glue preservative, chemical intermediate, anti-mildew treatment, solvent for polyester fiber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trichlorophenol, antiseptic, defoliant, and glue preservative, chemical intermediate, anti-mildew treatment, solvent for polyester fiber.

Solubility
Soluble in water(0.8g/L).

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text