Thermo Scientific Chemicals

2,5-Dimethoxytetrahydrofuran, cis + trans, 98%, Thermo Scientific Chemicals

Catalog number: A11687.36
500 g, Each
Thermo Scientific Chemicals

2,5-Dimethoxytetrahydrofuran, cis + trans, 98%, Thermo Scientific Chemicals

Catalog number: A11687.36
500 g, Each
Quantity
Catalog number: A11687.36
also known as A11687-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
696-59-3
IUPAC Name
2,5-dimethoxyoxolane
Molecular Formula
C6H12O3
InChI Key
GFISDBXSWQMOND-UHFFFAOYNA-N
SMILES
COC1CCC(OC)O1
Assay (GC)
>97.5% (sum of isomers)
Refractive Index
1.4160-1.4200 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Identification (FTIR)
Conforms
Form
Liquid

Description

2,5-Dimethoxytetrahydrofuran is a tetrahydrofuran derivative that finds application in proteomics research and in organic synthesis. It is also used as an intermediate for atropine sulfate, which is an anticholinergic drug. Further, it acts as a precursor of butanediol. In addition to this, it is used as a photographic hardener and as disinfectant in formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dimethoxytetrahydrofuran is a tetrahydrofuran derivative that finds application in proteomics research and in organic synthesis. It is also used as an intermediate for atropine sulfate, which is an anticholinergic drug. Further, it acts as a precursor of butanediol. In addition to this, it is used as a photographic hardener and as disinfectant in formulations.

Solubility
Miscible with methanol, diethyl ether and tetrahydrofuran. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text