Tin(II) oxalate, 98%, Thermo Scientific Chemicals
Tin(II) oxalate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tin(II) oxalate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2500 g
Catalog number A11689.36
also known as A11689-36
Price (USD)/ Each
75.65
Online exclusive
83.80 
Save 8.15 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
75.65
Online exclusive
83.80 
Save 8.15 (10%)
Add to cart
Tin(II) oxalate, 98%, Thermo Scientific Chemicals
Catalog numberA11689.36
Price (USD)/ Each
75.65
Online exclusive
83.80 
Save 8.15 (10%)
-
Add to cart
Chemical Identifiers
CAS814-94-8
SpecificationsSpecification SheetSpecification Sheet
Elemental AnalysisCu: <5ppm
Elemental AnalysisZn: <20ppm
Appearance (Color)White
Assay (special)≥97.5% (gravimetric assay)
Elemental AnalysisPb: <36ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only