Thermo Scientific™

2-Bromoanisole, 98%, Thermo Scientific Chemicals

Catalog number: A11692.14
25 g, Each
Thermo Scientific™

2-Bromoanisole, 98%, Thermo Scientific Chemicals

Catalog number: A11692.14
25 g, Each
Quantity
Catalog number: A11692.14
also known as A11692-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
578-57-4
IUPAC Name
1-bromo-2-methoxybenzene
Molecular Formula
C7H7BrO
InChI Key
HTDQSWDEWGSAMN-UHFFFAOYSA-N
SMILES
COC1=CC=CC=C1Br
Form
Liquid
Refractive Index
1.5730-1.5770 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%

Description

2-Bromoanisole was used in the synthesis of unsymmetrical substituted biphenyl compounds, used to produce (2-methoxy-phenyl)-acetone at the temperature of 100°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoanisole was used in the synthesis of unsymmetrical substituted biphenyl compounds, used to produce (2-methoxy-phenyl)-acetone at the temperature of 100°C.

Solubility
Soluble in ethanol and diethyl ether.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text