Thermo Scientific Chemicals

5-Nitroisophthalic acid, 97+%, Thermo Scientific Chemicals

Catalog number: A11696.22
100 g, Each
Thermo Scientific Chemicals

5-Nitroisophthalic acid, 97+%, Thermo Scientific Chemicals

Catalog number: A11696.22
100 g, Each
Quantity
Catalog number: A11696.22
also known as A11696-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
618-88-2
IUPAC Name
5-nitrobenzene-1,3-dicarboxylic acid
Molecular Formula
C8H5NO6
InChI Key
NBDAHKQJXVLAID-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC(=CC(=C1)C(O)=O)[N+]([O-])=O
Identification (FTIR)
Conforms
Melting Point (clear melt)
258.0-265.0°C
Appearance (Color)
White to pale yellow
Form
Powder
Assay (Aqueous acid-base Titration)
≥97.0 to ≤103.0%

Description

5-Nitroisophthalic acid is used as an intermediate of medicine compound and as a disperser in dyes. It is used as an intermediate in organic synthesis,agrochemical,pharmaceutical and dyestuffs etc,.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Nitroisophthalic acid is used as an intermediate of medicine compound and as a disperser in dyes. It is used as an intermediate in organic synthesis,agrochemical,pharmaceutical and dyestuffs etc,.

Solubility
Soluble in water.1.5 g/L at 20°C

Notes
Incompatible with strong oxidizing agents, strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text