Thermo Scientific Chemicals

Ethanolamine, 98+%, Thermo Scientific Chemicals

Catalog number: A11697.0E
2500 g, Each
Thermo Scientific Chemicals

Ethanolamine, 98+%, Thermo Scientific Chemicals

Catalog number: A11697.0E
2500 g, Each
Quantity
Catalog number: A11697.0E
also known as A11697-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
141-43-5
IUPAC Name
2-aminoethan-1-ol
Molecular Formula
C2H7NO
InChI Key
HZAXFHJVJLSVMW-UHFFFAOYSA-N
SMILES
NCCO
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥98.0%
Refractive Index
1.4525-1.4565 @ 20?C
Water Content (Karl Fischer Titration)
≤1%

Description

Ethanolamine is also used in messenger molecules such as palmitoylethanolamide, which have an effect on CB1 receptors. Ethanolamine is used as a scrubber for the removal of methyl bromide. Polyaniline doping ethanolamine is used for modified electrode preparation to measure uric acid in human body with the presence of antiscorbutic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethanolamine is also used in messenger molecules such as palmitoylethanolamide, which have an effect on CB1 receptors. Ethanolamine is used as a scrubber for the removal of methyl bromide. Polyaniline doping ethanolamine is used for modified electrode preparation to measure uric acid in human body with the presence of antiscorbutic acid.

Solubility
Miscible with water.

Notes
Store it away from heat, spark, open flames, hot surfaces and smoke. The storage area should be safe and enclosed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text