Thermo Scientific Chemicals

2-Chloro-2-methylbutane, 95%, Thermo Scientific Chemicals

Catalog number: A11703.22
100 g, Each
Thermo Scientific Chemicals

2-Chloro-2-methylbutane, 95%, Thermo Scientific Chemicals

Catalog number: A11703.22
100 g, Each
Quantity
Catalog number: A11703.22
also known as A11703-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
594-36-5
IUPAC Name
2-chloro-2-methylbutane
Molecular Formula
C5H11Cl
InChI Key
CRNIHJHMEQZAAS-UHFFFAOYSA-N
SMILES
CCC(C)(C)Cl
Form
Liquid
Refractive Index
1.4015-1.4075 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥94.0%

Description

2-Chloro-2-methylbutane is used as a solvent for the synthesis of other compounds and pentane. It is used in organic synthesis, Catalytic agent, Petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-2-methylbutane is used as a solvent for the synthesis of other compounds and pentane. It is used in organic synthesis, Catalytic agent, Petrochemical additive.

Solubility
Soluble in ethanol, ethyl ether. Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents. Avoid extremes of temperature and direct sunlight.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text