1-Fluoro-3-iodobenzene, 99%, Thermo Scientific Chemicals
1-Fluoro-3-iodobenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Fluoro-3-iodobenzene, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number A11706.22
also known as A11706-22
Price (USD)/ Each
279.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
279.00
Add to cart
1-Fluoro-3-iodobenzene, 99%, Thermo Scientific Chemicals
Catalog numberA11706.22
Price (USD)/ Each
279.00
-
Add to cart
Chemical Identifiers
CAS1121-86-4
IUPAC Name1-fluoro-3-iodobenzene
Molecular FormulaC6H4FI
InChI KeyVSKSBSORLCDRHS-UHFFFAOYSA-N
SMILESFC1=CC=CC(I)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
StabilizerCopper powder
Appearance (Color)Clear colorless to yellow
Assay (GC)≥98.5%
Refractive Index1.5825-1.5860 @ 20?C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only