Thermo Scientific Chemicals

Diethyl acetylenedicarboxylate, 96%, Thermo Scientific Chemicals

Catalog number: A11719.22
100 g, Each
Thermo Scientific Chemicals

Diethyl acetylenedicarboxylate, 96%, Thermo Scientific Chemicals

Catalog number: A11719.22
100 g, Each
Quantity
Catalog number: A11719.22
also known as A11719-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
762-21-0
IUPAC Name
1,4-diethyl but-2-ynedioate
Molecular Formula
C8H10O4
InChI Key
STRNXFOUBFLVIN-UHFFFAOYSA-N
SMILES
CCOC(=O)C#CC(=O)OCC
Refractive Index
1.4405-1.4465 @ 20?C
Form
Liquid
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥95.0%

Description

Diethyl acetylenedicarboxylate is used as a protein cross-linker. It is also used in the synthesis of 3,4,5-trisubstituted 2(5H)-furanone derivatives, highly functionalized thiazolidinone derivatives, novel cyclic peroxide glucosides and 4,11-dimesitylbisanthene, soluble bisanthene derivative, via Diels-Alder reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl acetylenedicarboxylate is used as a protein cross-linker. It is also used in the synthesis of 3,4,5-trisubstituted 2(5H)-furanone derivatives, highly functionalized thiazolidinone derivatives, novel cyclic peroxide glucosides and 4,11-dimesitylbisanthene, soluble bisanthene derivative, via Diels-Alder reaction.

Solubility
Soluble in ethanol, ethyl ether, CCl4, Insoluble in water.

Notes
Store at 4°C. Protect from heat. Store away from oxidizing agents. Incompatible with strong acids and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text