Thermo Scientific Chemicals

Diphenylphosphinic chloride, 97+%, Thermo Scientific Chemicals

Catalog number: A11721.22
100 g, Each
Thermo Scientific Chemicals

Diphenylphosphinic chloride, 97+%, Thermo Scientific Chemicals

Catalog number: A11721.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A11721.22
also known as A11721-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1499-21-4
IUPAC Name
diphenylphosphinoyl chloride
Molecular Formula
C12H10ClOP
InChI Key
QPQGTZMAQRXCJW-UHFFFAOYSA-N
SMILES
ClP(=O)(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
Clear colorless to yellow
Refractive Index
1.6065-1.6125 @ 20?C
Form
Liquid
Assay (GC)
≥97.0%

Description

Diphenylphosphinic chloride is a reagent that is used in the synthesis of bidentate ligands and peptide coupling agents. It is used as flame retardants. It also act as pharmaceutical adjuvant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenylphosphinic chloride is a reagent that is used in the synthesis of bidentate ligands and peptide coupling agents. It is used as flame retardants. It also act as pharmaceutical adjuvant.

Solubility
Decomposes with water.

Notes
Moisture Sensitive. Store away from strong bases, moisture. Incompatible with oxidizing agents,, Amines, Alcohols.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text