It may be a useful urinary biomarker for certain cancers such as gastric cancer and oxidative stress.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It may be a useful urinary biomarker for certain cancers such as gastric cancer and oxidative stress.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
Luisa M Ostertag et. al. In vitro anti-platelet effects of simple plant-derived phenolic compounds are only found at high, non-physiological concentrations. Molecular Nutrition & Food Research. 2011, 55 (11), 1624-1636.
Thomas Frank; Gabriele Netzel; Dietmar R Kammerer; Reinhold Carle; Adolf Kler; Erwin Kriesl; Irmgard Bitsch; Roland Bitsch; Michael Netzel. Consumption of Hibiscus sabdariffa L. aqueous extract and its impact on systemic antioxidant potential in healthy subjects. Journal of the Science of Food and Agriculture. 2012, 92 (10), 2207-2218.