Thermo Scientific Chemicals

Hydantoin-5-acetic acid, 98%, Thermo Scientific Chemicals

Catalog number: A11731.06
5 g, Each
Thermo Scientific Chemicals

Hydantoin-5-acetic acid, 98%, Thermo Scientific Chemicals

Catalog number: A11731.06
5 g, Each
Quantity
Catalog number: A11731.06
also known as A11731-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5427-26-9
IUPAC Name
2-(2,5-dioxoimidazolidin-4-yl)acetic acid
Molecular Formula
C5H6N2O4
InChI Key
DQQLZADYSWBCOX-UHFFFAOYNA-N
SMILES
OC(=O)CC1NC(=O)NC1=O
Form
Crystalline powder
Melting Point (clear melt)
ca. 210-225?C dec.
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text