3'-Fluoroacetophenone is used as raw material and intermediate in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3′-Fluoroacetophenone is used as raw material and intermediate in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
Solubility
Slightly soluble in water.
Notes
Incompatible with strong bases, strong oxidizing agents, strong reducing agents. Ensure adequate ventilation during use. Use only in a chemical fume hood.
RUO – Research Use Only
General References:
- Jan von Langermann; Annett Mell;, Eckhard Paetzold; Thomas Daußmann and Udo Kragl. Hydroxynitrile Lyase in Organic Solvent-Free Systems to Overcome Thermodynamic Limitations.Advanced Synthesis & Catalysis. 2007, 349 1418-1424.
- Asgeir Bjarnason; James W. Taylor; James A. Kinsinger; Robert B. Cody; David A. Weil. Isomer discrimination of disubstituted benzene derivatives through gas-phase iron(I) ion reactions in a Fourier-transform mass spectrometer.Anal. Chem. 1989, 61 (17), 1889-1894.