4-Biphenylacetic acid, 98%, Thermo Scientific Chemicals
4-Biphenylacetic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Biphenylacetic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number A11749.30
also known as A11749-30
Price (USD)/ Each
741.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
741.00
Add to cart
4-Biphenylacetic acid, 98%, Thermo Scientific Chemicals
Catalog numberA11749.30
Price (USD)/ Each
741.00
-
Add to cart
Chemical Identifiers
CAS5728-52-9
IUPAC Name2-{[1,1'-biphenyl]-4-yl}acetic acid
Molecular FormulaC14H12O2
InChI KeyQRZAKQDHEVVFRX-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC=C(C=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream to pale brown
Melting Point (clear melt)158.0-165.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only