1-Chloro-4-phenylbutane, 97%, Thermo Scientific Chemicals
1-Chloro-4-phenylbutane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Chloro-4-phenylbutane, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number A11753.14
also known as A11753-14
Price (USD)/ Each
314.65
Online exclusive
349.00 
Save 34.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
314.65
Online exclusive
349.00 
Save 34.35 (10%)
Add to cart
1-Chloro-4-phenylbutane, 97%, Thermo Scientific Chemicals
Catalog numberA11753.14
Price (USD)/ Each
314.65
Online exclusive
349.00 
Save 34.35 (10%)
-
Add to cart
Chemical Identifiers
CAS4830-93-7
IUPAC Name(4-chlorobutyl)benzene
Molecular FormulaC10H13Cl
InChI KeyFLLZCZIHURYEQP-UHFFFAOYSA-N
SMILESClCCCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5160-1.5210 @ 20?C
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only