Thermo Scientific Chemicals

delta-Decanolactone, 97%, Thermo Scientific Chemicals

Catalog number: A11756.22
100 g, Each
Thermo Scientific Chemicals

delta-Decanolactone, 97%, Thermo Scientific Chemicals

Catalog number: A11756.22
100 g, Each
Quantity
Catalog number: A11756.22
also known as A11756-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
705-86-2
IUPAC Name
6-pentyloxan-2-one
Molecular Formula
C10H18O2
InChI Key
GHBSPIPJMLAMEP-UHFFFAOYNA-N
SMILES
CCCCCC1CCCC(=O)O1
Appearance (Color)
Clear colorless
Refractive Index
1.4555-1.4605 @ 20?C
Form
Liquid
Assay (GC)
≥96.0%

Description

Used in pharmaceutical preparations, feed additive, flavorings agent. Also used by food and flavor industries for its coconut fragrance and taste. It is also used as a reagent in the synthesis of (±)-Massoilactone (M197600); a chemical component with antibacterial activity found in volatile oils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in pharmaceutical preparations, feed additive, flavorings agent. Also used by food and flavor industries for its coconut fragrance and taste. It is also used as a reagent in the synthesis of (±)-Massoilactone (M197600); a chemical component with antibacterial activity found in volatile oils.

Solubility
Soluble in water (4 mg/ml at 28°C), alcohols, and propylene glycol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text