Thermo Scientific Chemicals

4-Bromo-3,5-dimethyl-1H-pyrazole, 98%, Thermo Scientific Chemicals

Catalog number: A11759.14
25 g, Each
Thermo Scientific Chemicals

4-Bromo-3,5-dimethyl-1H-pyrazole, 98%, Thermo Scientific Chemicals

Catalog number: A11759.14
25 g, Each
Quantity
Catalog number: A11759.14
also known as A11759-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3398-16-1
IUPAC Name
4-bromo-3,5-dimethyl-1H-pyrazole
Molecular Formula
C5H7BrN2
InChI Key
RISOHYOEPYWKOB-UHFFFAOYSA-N
SMILES
CC1=C(Br)C(C)=NN1
Appearance (Color)
White to pale cream or pale yellow
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
117-124°C
Form
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text