Thermo Scientific Chemicals

Allyl bromide, 99%, stab. with 300-1000ppm Propylene oxide, Thermo Scientific Chemicals

Catalog number: A11766.30
250 g, Each
Thermo Scientific Chemicals

Allyl bromide, 99%, stab. with 300-1000ppm Propylene oxide, Thermo Scientific Chemicals

Catalog number: A11766.30
250 g, Each
Quantity
Catalog number: A11766.30
also known as A11766-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
106-95-6
IUPAC Name
3-bromoprop-1-ene
Molecular Formula
C3H5Br
InChI Key
BHELZAPQIKSEDF-UHFFFAOYSA-N
SMILES
BrCC=C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥98.5%
Refractive Index
1.4670-1.4710 @ 20°C
Stabilizer
Propylene oxide, 300-1,000ppm

Description

Allyl bromide is used as an alkylating agent in the synthesis of pharmaceuticals, polymers, adhesives, perfumes, biochemicals and other allylic compounds. It is used as precursor for the preparation of allyliczinc bromide by reacting it with zinc. It is also used in the preparation of allylethers like allyl decyl ether, allyl benzyl ether and allyl geranyl ether. It is also used in the preparation of R enantiomer of allyl phenyl carbinol (APC) such as 1-phenyl-3-butene, which is a valuable intermediate for drugs and agro-chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyl bromide is used as an alkylating agent in the synthesis of pharmaceuticals, polymers, adhesives, perfumes, biochemicals and other allylic compounds. It is used as precursor for the preparation of allyliczinc bromide by reacting it with zinc. It is also used in the preparation of allylethers like allyl decyl ether, allyl benzyl ether and allyl geranyl ether. It is also used in the preparation of R enantiomer of allyl phenyl carbinol (APC) such as 1-phenyl-3-butene, which is a valuable intermediate for drugs and agro-chemicals.

Solubility
Slightly miscible with water.

Notes
Moisture and light sensitive. Incompatible with oxidizing agents, alkali metals, alkaline earth metals, amides, amines and powdered metals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text