Thermo Scientific Chemicals

Trifluoromethanesulfonic anhydride, 98%, Thermo Scientific Chemicals

Catalog number: A11767.14
25 g, Each
Thermo Scientific Chemicals

Trifluoromethanesulfonic anhydride, 98%, Thermo Scientific Chemicals

Catalog number: A11767.14
25 g, Each
Quantity
Catalog number: A11767.14
also known as A11767-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
358-23-6
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay by NMR
≥97.5% (Fluorine NMR)
Comment
May contain slight sediment
Proton NMR
Conforms to structure

Description

Trifluoromethanesulfonic anhydride is used to convert phenols and imine into triflic ester and NTf group. It is a strong electrophile used for the introduction of triflyl group in chemical synthesis. It serves as a reagent in the preparation of alkyl and vinyl triflates, and for the stereoselective synthesis of mannosazide methyl uronate donors. It acts as a catalyst for glycosylation with anomeric hydroxy sugars to prepare polysaccharides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trifluoromethanesulfonic anhydride is used to convert phenols and imine into triflic ester and NTf group. It is a strong electrophile used for the introduction of triflyl group in chemical synthesis. It serves as a reagent in the preparation of alkyl and vinyl triflates, and for the stereoselective synthesis of mannosazide methyl uronate donors. It acts as a catalyst for glycosylation with anomeric hydroxy sugars to prepare polysaccharides.

Solubility
Miscible with dichloromethane. Immiscible with hydrocarbons.

Notes
Moisture sensitive. Incompatible with water, strong bases, strong oxidizing agents, acids and alcohols.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text