Trifluoromethanesulfonic anhydride, 98%, Thermo Scientific Chemicals
Trifluoromethanesulfonic anhydride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trifluoromethanesulfonic anhydride, 98%, Thermo Scientific Chemicals

Alfa Aesar Trifluoromethanesulfonic anhydride, 98%, Quantity: 250g, Boiling Point: 81 deg.C to 83 deg.C, Melting Point: -80 deg.C, Molecular Weight: 282.127, Percent Purity: 98%, Beilstein: 1813600, CAS: 358-23-6, ChEBI: CHEBI:48509, Density: 1.72
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
25 g
50 g
100 g
Catalog number A11767.30
Price (USD)/ Each
621.65
Online exclusive
690.00 
Save 68.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
621.65
Online exclusive
690.00 
Save 68.35 (10%)
Add to cart
Trifluoromethanesulfonic anhydride, 98%, Thermo Scientific Chemicals
Catalog numberA11767.30
Price (USD)/ Each
621.65
Online exclusive
690.00 
Save 68.35 (10%)
-
Add to cart
Chemical Identifiers
CAS358-23-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay by NMR≥97.5% (Fluorine NMR)
CommentMay contain slight sediment
Proton NMRConforms to structure
View more
Trifluoromethanesulfonic anhydride is used to convert phenols and imine into triflic ester and NTf group. It is a strong electrophile used for the introduction of triflyl group in chemical synthesis. It serves as a reagent in the preparation of alkyl and vinyl triflates, and for the stereoselective synthesis of mannosazide methyl uronate donors. It acts as a catalyst for glycosylation with anomeric hydroxy sugars to prepare polysaccharides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trifluoromethanesulfonic anhydride is used to convert phenols and imine into triflic ester and NTf group. It is a strong electrophile used for the introduction of triflyl group in chemical synthesis. It serves as a reagent in the preparation of alkyl and vinyl triflates, and for the stereoselective synthesis of mannosazide methyl uronate donors. It acts as a catalyst for glycosylation with anomeric hydroxy sugars to prepare polysaccharides.

Solubility
Miscible with dichloromethane. Immiscible with hydrocarbons.

Notes
Moisture sensitive. Incompatible with water, strong bases, strong oxidizing agents, acids and alcohols.
RUO – Research Use Only

General References:

  1. For examples of preparation of alkyl and aryl triflates, and use of the latter in the Stille coupling reaction, see: Org. Synth. Coll., 6, 324 (1988); 9, 553 (1998). Alkyl triflates undergo solvolysis reactions between five and seven powers of ten times more rapidly than the corresponding halides or tosylates: J. Am. Chem. Soc., 97, 6478 (1975); Angew. Chem. Int. Ed., 9, 521 (1970). For a review of the chemistry of triflate esters, see: Synthesis, 85 (1982).
  2. Enolizable carbonyl compounds in the presence of base can be converted to vinyl (enol) triflates: Synthesis, 85 (1982); Org. Synth. Coll., 6, 757 (1988). For conditions employing preferred base, 2,6-Di-tert-butyl-4-methyl pyridine, L14143, see: Org. Synth. Coll., 8, 97, 126 (1993). Enol triflates behave as a source of vinyl cations. For reviews, see: Angew. Chem. Int. Ed., 17, 333 (1978); Acc. Chem. Res., 11, 107 (1978); 15, 348 (1982); 21, 147 (1988).
  3. For formation of keteniminium triflates, see N,N-Dimethyl acetamide, A10924.
  4. The Ritter reaction is normally most successful with tertiary alcohols. In contrast, a useful variation allows conversion of primary or secondary alcohols to amides in good yield: Tetrahedron Lett., 30, 581 (1989):
  5. For use in the Vilsmeier formylation of less active substrates, see N,N-Dimethyl formamide, A13547.
  6. Triflic anhydride has also been found to promote the nitration of arenes, even deactivated ones, under mild conditions: Synthesis, 1087 (1992).
  7. For a review of chemical transformations induced by the reagent, see: Tetrahedron, 56, 3077 (2000). For a brief feature on uses in synthesis, see: Synlett, 390 (2004).