Mesitylene-2-sulfonyl chloride, 99%, Thermo Scientific Chemicals
Mesitylene-2-sulfonyl chloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Mesitylene-2-sulfonyl chloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A11775.14
also known as A11775-14
Price (USD)/ Each
63.60
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
63.60
Add to cart
Mesitylene-2-sulfonyl chloride, 99%, Thermo Scientific Chemicals
Catalog numberA11775.14
Price (USD)/ Each
63.60
-
Add to cart
Chemical Identifiers
CAS773-64-8
IUPAC Name2,4,6-trimethylbenzene-1-sulfonyl chloride
Molecular FormulaC9H11ClO2S
InChI KeyPVJZBZSCGJAWNG-UHFFFAOYSA-N
SMILESCC1=CC(C)=C(C(C)=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Melting Point (clear melt)50.0-56.0?C
Mesitylene-2-sulfonyl chloride is used as reagent for intramolecular lactamizations and lactonization. It also acts as a reagent for protecting guanidino groups and tryptophan. It is used as coupling reagent in polynucleotide synthesis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mesitylene-2-sulfonyl chloride is used as reagent for intramolecular lactamizations and lactonization. It also acts as a reagent for protecting guanidino groups and tryptophan. It is used as coupling reagent in polynucleotide synthesis

Solubility
Soluble in toluene (almost transparency).

Notes
Moisture Sensitive. Store under inert gas. Incompatible with strong bases. Store away from moisture.
RUO – Research Use Only

General References:

  1. Chi-Hua Wang; Saul G. Cohen. Mercaptan Catalysis in Thermoneutral Free Radical Exchange.J. Am. Chem. Soc. 1957, 79 (8),1924-1929.
  2. Adeleh Kiania; Batool Akhlaghiniaa; Hamed Rouhi-Saadabada & Mehdi Bakavolia.Journal of Sulfur Chemistry. 2014, 35 119-127.
  3. Selective reagent for sulfonylation of carbohydrates: J. Chem. Soc., Perkin 1, 1373 (1974).
  4. For reviews of the use of hindered sulfonyl chlorides as condensing agents for the formation of the phosphate link of oligonucleotides, see: Angew. Chem. Int. Ed., 11, 451 (1972); Synthesis, 222 (1975); Chem. Rev., 77, 183 (1977). For reviews of oligonucleotide synthesis by the phosphotriester method, see: Heterocycles, 7, 1197 (1977); Tetrahedron, 34, 3143 (1978). Compare 2,4,6-Triisopropyl benzenesulfonyl chloride, A11458.
  5. Reagent for protection of the guanidino function of arginine residues as mesitylenesulfonyl (Mts) derivatives in peptide synthesis: J. Chem. Soc., Chem. Commun., 482 (1978); Chem. Pharm. Bull., 26, 3752 (1978); Int. J. Pept. Prot. Res., 14, 169 (1979). Deprotection can be effected with triflic acid in TFA in the presence of thioanisole as a cation scavenger: J. Chem. Soc., Chem. Commun., 1063 (1980).
  6. The indole nitrogen, e.g. in tryptophan residues, may also be protected as the Mts amide, stable to dilute alkali, hydrazine hydrate, TFA, 4N HCl in dioxane, or HBr in AcOH, but cleaved by MsOH, or TfOH in TFA: Chem. Pharm. Bull., 32, 2660 (1984). See Appendix 6.