Thermo Scientific Chemicals

Mesitylene-2-sulfonyl chloride, 99%, Thermo Scientific Chemicals

Catalog number: A11775.22
100 g, Each
Thermo Scientific Chemicals

Mesitylene-2-sulfonyl chloride, 99%, Thermo Scientific Chemicals

Catalog number: A11775.22
100 g, Each
Quantity
Catalog number: A11775.22
also known as A11775-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
773-64-8
IUPAC Name
2,4,6-trimethylbenzene-1-sulfonyl chloride
Molecular Formula
C9H11ClO2S
InChI Key
PVJZBZSCGJAWNG-UHFFFAOYSA-N
SMILES
CC1=CC(C)=C(C(C)=C1)S(Cl)(=O)=O
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
50.0-56.0?C

Description

Mesitylene-2-sulfonyl chloride is used as reagent for intramolecular lactamizations and lactonization. It also acts as a reagent for protecting guanidino groups and tryptophan. It is used as coupling reagent in polynucleotide synthesis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mesitylene-2-sulfonyl chloride is used as reagent for intramolecular lactamizations and lactonization. It also acts as a reagent for protecting guanidino groups and tryptophan. It is used as coupling reagent in polynucleotide synthesis

Solubility
Soluble in toluene (almost transparency).

Notes
Moisture Sensitive. Store under inert gas. Incompatible with strong bases. Store away from moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text