Thermo Scientific Chemicals

Tetraethylammonium iodide, 98+%, Thermo Scientific Chemicals

Catalog number: A11783.0B
1000 g, Each
Thermo Scientific Chemicals

Tetraethylammonium iodide, 98+%, Thermo Scientific Chemicals

Catalog number: A11783.0B
1000 g, Each
Quantity
Catalog number: A11783.0B
also known as A11783-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
68-05-3
Appearance (Color)
White
Form
Crystalline powder or crystals
Assay (Titration ex Iodide)
≥98.0 to ≤102.0% (non-U.S. specification)
Assay from Suppliers CofA
≥98.0% (U.S specification)
Comment
Specification differs for U.S. and non-U.S. material where indicated

Description

It has been used as the source of tetraethylammonium ions in pharmacological and physiological studies, but is also used in organic chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has been used as the source of tetraethylammonium ions in pharmacological and physiological studies, but is also used in organic chemical synthesis.

Solubility
Soluble in water almost transparency.

Notes
Light Sensitive and Hygroscopic. Store in dark and away from water/moisture. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text