Thermo Scientific Chemicals

2-Bromoaniline, 98%, Thermo Scientific Chemicals

Catalog number: A11788.14
25 g, Each
Thermo Scientific Chemicals

2-Bromoaniline, 98%, Thermo Scientific Chemicals

Catalog number: A11788.14
25 g, Each
Quantity
Catalog number: A11788.14
also known as A11788-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
615-36-1
IUPAC Name
2-bromoaniline
Molecular Formula
C6H6BrN
InChI Key
AOPBDRUWRLBSDB-UHFFFAOYSA-N
SMILES
NC1=CC=CC=C1Br
Appearance (Color)
White to orange or brown
Form
Fused solid (clear liquid as melt)
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms

Description

2-Bromoaniline is used as a corrosion inhibitor, emulsifying and antiseptic agents. It is used in the rubber industry, e.g. diphenylguanidines, phenylenediamines mercaptobenzothiazoles, aniline ketones and etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoaniline is used as a corrosion inhibitor, emulsifying and antiseptic agents. It is used in the rubber industry, e.g. diphenylguanidines, phenylenediamines mercaptobenzothiazoles, aniline ketones and etc.

Solubility
Insoluble in water.

Notes
Store at 2-8°C. Incompatible with acids, Acid chlorides, Acid anhydrides, Chloroformates, Strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text