2-Bromoaniline is used as a corrosion inhibitor, emulsifying and antiseptic agents. It is used in the rubber industry, e.g. diphenylguanidines, phenylenediamines mercaptobenzothiazoles, aniline ketones and etc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromoaniline is used as a corrosion inhibitor, emulsifying and antiseptic agents. It is used in the rubber industry, e.g. diphenylguanidines, phenylenediamines mercaptobenzothiazoles, aniline ketones and etc.
Solubility
Insoluble in water.
Notes
Store at 2-8°C. Incompatible with acids, Acid chlorides, Acid anhydrides, Chloroformates, Strong oxidizing agents.
RUO – Research Use Only
Michael E. Krolski; Alfred F. Renaldo; Duane E. Rudisill; J. K. Stille. Palladium-catalyzed coupling of 2-bromoanilines with vinylstannanes. A regiocontrolled synthesis of substituted indoles.J. Org. Chem. 1988, 53 (6), 1170-1176.
Khaled Othmen; Pierre Boule. Photochemical behaviour of 2-halogenoanilines in aqueous solution. Journal of Photochemistry and Photobiology A: Chemistry. 2000, 136 79-86.