2,4-Dichlorophenylacetic acid, 98+%, Thermo Scientific Chemicals
2,4-Dichlorophenylacetic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4-Dichlorophenylacetic acid, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number A11791.14
also known as A11791-14
Price (USD)/ Each
69.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
69.00
Add to cart
2,4-Dichlorophenylacetic acid, 98+%, Thermo Scientific Chemicals
Catalog numberA11791.14
Price (USD)/ Each
69.00
-
Add to cart
Chemical Identifiers
CAS19719-28-9
IUPAC Name2-(2,4-dichlorophenyl)acetic acid
Molecular FormulaC8H6Cl2O2
InChI KeyGXMWLJKTGBZMBH-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC=C(Cl)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder or lumps
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Melting Point (clear melt)126.0-133.0?C
Assay (Silylated GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only