Thermo Scientific Chemicals

3-Methyl-1-phenyl-2-phospholene 1-oxide, 94%, Thermo Scientific Chemicals

Catalog number: A11792.22
100 g, Each
Thermo Scientific Chemicals

3-Methyl-1-phenyl-2-phospholene 1-oxide, 94%, Thermo Scientific Chemicals

Catalog number: A11792.22
100 g, Each
Quantity
Catalog number: A11792.22
also known as A11792-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
707-61-9
IUPAC Name
4-methyl-1-phenyl-2,3-dihydro-1H-1λ⁵-phosphol-1-one
Molecular Formula
C11H13OP
InChI Key
YMKWWHFRGALXLE-UHFFFAOYNA-N
SMILES
CC1=CP(=O)(CC1)C1=CC=CC=C1
Melting Point (clear melt)
57-66°C
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
White to cream
Assay (GC)
≥92.0%

Description

3-Methyl-1-phenyl-2-phospholene 1-oxide is used as a catalyst for intramolecular aza-wittig cyclization reactions and polymerization reactions. It is a reactant used for the preparation of phospha sugars and their radical bromination derivatives as anticancer agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-1-phenyl-2-phospholene 1-oxide is used as a catalyst for intramolecular aza-wittig cyclization reactions and polymerization reactions. It is a reactant used for the preparation of phospha sugars and their radical bromination derivatives as anticancer agents.

Solubility
Soluble in polar solvents.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text