Isonipecotic acid, 98%, Thermo Scientific Chemicals
Isonipecotic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Isonipecotic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A11795.14
also known as A11795-14
Price (USD)/ Each
55.10
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
55.10
Add to cart
Isonipecotic acid, 98%, Thermo Scientific Chemicals
Catalog numberA11795.14
Price (USD)/ Each
55.10
-
Add to cart
Chemical Identifiers
CAS498-94-2
IUPAC Namepiperidine-4-carboxylic acid
Molecular FormulaC6H11NO2
InChI KeySRJOCJYGOFTFLH-UHFFFAOYSA-N
SMILESOC(=O)C1CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
FormPowder
Assay (Silylated GC)≥97.5%
A GABA-A receptor partial agonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only